Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
1010-02
100242-266EA 129.46 USD
100242-266
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-Mouse Ig-FITC antibody with minimal reactivity to human proteins for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR