Anti-Ig Goat Antibody

Supplier: Southern Biotechnology
1010-01
100242-264EA 151.69 USD
100242-264
Anti-Ig Goat Antibody
Antibodies
Purified Goat Anti-Mouse Ig antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR