SP Wilmad-LabGlass Bent and Straight Hose Connections, SP Industries
Supplier: SP Industries
LG-10818-104 LG-10818-102
14007-638EA 22.69 USD
14007-638 14007-636
SP Wilmad-LabGlass Bent and Straight Hose Connections, SP Industries
Fittings and Connectors
Borosilicate glass hose connecting units are used to connect to an apparatus with GL14 or GL18 thread sizes
Complete connecting unit consists of connector (either straight or bent) and a cap with hole in center.