Methanol ≥99.9% ACS, Ph. Eur., super gradient grade for HPLC, suitable for UPLC/UHPLC instruments, VWR Chemicals BDH®

Danger

Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

BDH85681.400 BDHVBDH85681.050 BDHVBDH85681.201 BDHVBDH85681.202
BDH85681.400CS 354.78 USD
BDH85681.400 BDH85681.202 BDH85681.201 BDH85681.050
Methanol ≥99.9% ACS, Ph. Eur., super gradient grade for HPLC, suitable for UPLC/UHPLC instruments, VWR Chemicals BDH®
Methanol

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


Solvents are pumped through 0.2 micron filters on filling and packaged under Nitrogen.


Ordering information: To purchase these solvents in stainless steel returnable drums as part of the VWR ReCAP program, please contact your local VWR sales representative. A separate contract for VWR ReCAP products is required.


Packaging: All bottles have tamper evident caps for added peace of mind and are packed in environmentally –friendly corrugated/pulp case inserts that are fully recyclable.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64…65 °C
Density: 0.791…0.793
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Specification Test Results

Assay (GC) Min. 99.9%
Carbonyl compounds Passes Test
Identification Passes Test
Solubility in water Passes Test
Substances darkened by sulfuric acid Passes Test
Substances reducing permanganate Passes Test
Acidity Max. 0.0002 mEq/g
Alkalinity Max. 0.0002 mEq/g
Coloration Max. 10 APHA
Density (20/20) 0.791 - 0.793
Evaporation residue Max. 1 ppm
Water Max. 0.02%
Gradient (235 nm) Max. 2 mAU
Gradient (254 nm) Max. 1 mAU
Filtered at 0.2 μm Confirmed
Transmittance (210 nm) Min. 45%
Transmittance (220 nm) Min. 65%
Transmittance (225 nm) Min. 70%
Transmittance (230 nm) Min. 83%
Transmittance (235 nm) Min. 85%
Transmittance (240 nm) Min. 90%
Transmittance (250 nm) Min. 95%
Transmittance (260 nm) Min. 98%
Transmittance (280 - 400 nm) Min. 98%
Absorbance (210 nm) Max. 0.347
Absorbance (220 nm) Max. 0.188
Absorbance (225 nm) Max. 0.155
Absorbance (230 nm) Max. 0.081
Absorbance (235 nm) Max. 0.071
Absorbance (240 nm) Max. 0.046
Absorbance (250 nm) Max. 0.023
Absorbance (260 nm) Max. 0.009
Absorbance (280 - 400 nm) Max. 0.009
Fluorescence (as quinine) (254 nm) Max. 1 ppb
Fluorescence (as quinine) (365 nm) Max. 0.5 ppb
Conforms Ph.Eur. R1 1053201 Passes Test
Conforms Ph.Eur. R2 1053202 Passes Test
Conforms to ACS Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR