SHOWA 250 Kevlar/Steel Cut Resistant Glove, Showa
Supplier: Showa
250-09 250-11 250-08 250-07
89208-292DZ 459.11 USD
89208-292 89208-286 89208-288 89208-284
SHOWA 250 Kevlar/Steel Cut Resistant Glove, Showa
Gloves
High level Ansi 4 cut protection with breathable Kevlar/Steel liner.
  • Kevlar/Steel liner provides high level protection
  • Sponge Nitrile coating for superior grip in oily conditions
Extreme protection from cuts and an optimum oil grip. Coating provides protection from sharp edges and withstands abrasions, snags, and punctures.
Certifications: Meets ASTM standards EN-388.
Ordering information: Ordered by case of 72 pair.
Packaging: Packaged 6 poly bags of 12 pair per Master Case.