Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microwave Oven, 2.2 Cu. Ft., Argos Technologies

Supplier: ARGOS TECHNOLOGIES INC.
111092
Offers Inverter Technology for true and consistent distribution of microwave energy.

  • Membrane keypad
  • Inverter turbo defrost technology
  • 10 power levels
  • Keep warm mode
  • Delay start
  • Timer
  • Quick minute

This 1250 W, 2.2CF oven offers Inverter Technology for true and consistent distribution of microwave energy

Other highlights include a Membrane keypad, Inverter Turbo Defrost technology, 10 Power levels, Keep Warm mode, Delay Start, Timer, Quick Minute, 16.5” Turntable and More/Less Control. White Finish.

Conventional microwaves operate on only one power level; the microwave energy is either on or off. Inverter technology delivers true low, medium and high power levels which help prevent cold spots and overheated edges and surfaces. The large 61.1 L interior accommodates 1 L media bottles and 2 L Griffin flasks.

Certifications: UL listed.

Caution: Note: This unit is not explosion-proof and is not intended for use in acid digestion applications.

Upper-left corner of door has a small indentation to allow smooth opening of door.



 

ORDER

Expand All / Collapse All
Description Volume Exterior Length Inside Height Inside Width Inner Length Outside Height Outside Width Supplier No. VWR Catalog Number Unit Price Quantity
Microwave Oven 2.2 Cu. Ft. 23 7/8 10 15/16 18 7/16 18 1/2 14” 19 7/16 111092 75856-526 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Electrical 1250W, 120V 50/60Hz

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty