Air Mass Generator, For 10 Groups ESCP
3215
470000-050KT 307.99 USD
470000-050
Air Mass Generator, For 10 Groups ESCP
Investigate how air mass is formed and how temperature relates to different types of air masses. Two add-ons at different heights inside a tube show how temperature varies with altitude. The set includes one 2-1/2"x 18" transparent and markable tube with integral thermometers on a track, two-piece convection chamber, tubing, funnel, smoke generator and Teacher's Guide.