Pyrene N-hexadecylsulfonamide, Marker Gene Technologies, Inc.
M0525
101447-504EA 148.15 USD
101447-504
Pyrene N-hexadecylsulfonamide, Marker Gene Technologies, Inc.
Polar, neutral, blue-fluorescent probe for labeling of membranes in vivo.
  • Lipid probe
This live cell probe has also been used for analysis of drug delivery mechanisms.