Agarose
Supplier: Promega
V3841
PAV3841EA 176.62 USD
PAV3841
Agarose
Enzymes
Premium agarose used for isolating DNA fragments from 10 to 1,000bp.
  • Quality tested and certified free of DNase and RNase activity
Agarose, LMP, Preparative Grade for Small Fragments (10 to 1,000bp), is a premium agarose used for isolating DNA fragments from 10 to 1,000bp. The isolated DNA fragments can be used in various molecular biology applications: 1) restriction digestion, 2) ligation and transformation, and 3) random prime labeling. LMP = low melting point (i.e., </=65 degrees C).
LMP stands for Low Melting Point.