PROMP106
P1062 P1061
PAP1062EA 3178.52 USD
PAP1062 PAP1061
PROMP106
Luciferin Substrate That is Cleaved by β-Galactosidase and Generates Light.
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings
Luciferin-β-galactoside is a substrate for the commonly used reporter enzyme β-galactosidase. The substrate is cleaved by β-galactosidase to form luciferin and galactose. When used in a model system expressing firefly luciferase, the luciferin is then utilized in a firefly luciferase reaction to generate light. Product integrity is ensured by packaging most products in amber vials with septa as well as offering ease of dilution and use for imaging experiments. VivoGlo™ Luciferin-β-Galactosidase Substrate is packaged with fine tolerances to minimize the need to weigh.
Storage: -30 °C to -10 °C