Coelenterazine h

Supplier: Promega Corporation

Synonyms: CLZN-h2,8-dibenzyl-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one2-Deoxycoelenterazine2-(4-Dehydroxy)coelenterazine

S2011
PAS2011EA 145.23 USD
PAS2011
Coelenterazine h
Coelenterazine h

We offer Coelenterazine (native) and Coelenterazine-h. Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin.


  • 95% pure
  • Supplied as a dried substrate for easy preparation in methanol or ethanol


Luciferases from Renilla, Aequorea and other marine organisms are commonly used as indicators or reporters for studying cellular phenomena in expression assays in eukaryotic cells. Renilla luciferase is often used as a reporter of transcription regulation, whereas apoaequorin is often used as a calcium indicator. Other uses of coelenterazines include chemiluminescent detection of Reactive Oxygen Species (ROS) in cells or tissues. We offer Coelenterazine (native) and Coelenterazine-h. Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin. Formula: C26H21N3O3. Formula Weight: 423.5. Form: Film. Coelenterazine-h imparts a luminescent intensity with its aequorin complex that is reported to be 10-20 times higher than that of native coelenterazine, making this derivative a useful tool for measuring small changes in Ca+ concentrations. Formula: C26H21N3O2. Formula Weight: 407.5. Form: Film.


Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin. Coelenterazine-h imparts a luminescent intensity with its aequorin complex that is reported to be 10–20 times higher than that of native coelenterazine, making this derivative a useful tool for measuring small changes in Ca²⁺ concentrations. Both Coelenterazine-h and Coelenterazine (native) are in film form.

Formula: C₂₆H₂₁N₃O₂
MW: 407.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798394
CAS Number: 50909-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR