- Home
- Assays
- Enzyme Assays
- HDAC-Glo I/II Screening System, Promega
HDAC-Glo I/II Screening System, Promega
Supplier: Promega
G6430
G6431
PAG6431EA
2455.51
USD
PAG6431
PAG6430
HDAC-Glo I/II Screening System, Promega
Assays
Enzyme Assays
The HDAC-Glo I/II Assays and Screening System are single-reagent-addition, homogeneous, luminescent assays that measure the relative activity of histone deacetylase (HDAC) class I and II enzymes from cells, extracts or purified enzyme sources.
- Detects HDAC Class I and II Enzyme Activity
- Simple add-mix-measure protocol
- Use purified enzymes, extracts or cells directly in culture plates
- Screening System includes HeLa Nuclear Extract
The HDAC-Glo I/II Assays and Screening System are single-reagent-addition, homogeneous, luminescent assays that measure the relative activity of histone deacetylase (HDAC) class I and II enzymes from cells, extracts or purified enzyme sources. The assays use an acetylated, live-cell-permeant, luminogenic peptide substrate that can be deacetylated by HDAC activities. Deacetylation of the peptide aminoluciferin substrate is measured using a coupled enzymatic system in which a protease in the Developer Reagent cleaves the deacetylated peptide from the aminoluciferin substrate, releasing aminoluciferin, which is quantified in a reaction using Ultra-Glo recombinant firefly luciferase. The assay reaction is typically complete within 15-45 minutes with no sample manipulation. The HDAC-mediated luminescent signal is persistent, with a half-life of greater than 3 hours, allowing batch processing of multiwell plates. The HDAC assay is broadly useful for class I and II enzymes. The Trichostatin A, included in the HDAC-Glo I/II Screening Systems or available separately, is a known pan HDAC inhibitor that may be used as a positive control inhibitor. The Trichostatin A is supplied at a concentration of 10mM in DMSO. The HeLa Nuclear Extract, included in the HDAC-Glo I/II Screening Systems or available separately, may be used as a source of histone deacetylase activity. The diluted extract also can be used as an HDAC-Glo I/II Assay chemistry control.
The HDAC-mediated luminescent signal is persistent, with a half-life of greater than three hours, allowing batch processing of multiwell plates. The HDAC assay is broadly useful for class I and II enzymes.
Deacetylation of the peptide aminoluciferin substrate is measured using a coupled enzymatic system in which a protease in the Developer Reagent cleaves the deacetylated peptide from the aminoluciferin substrate, releasing aminoluciferin, which is quantified in a reaction using Ultra-Glo™ recombinant firefly luciferase. The assay reaction is typically complete within 15–45 minutes with no sample manipulation.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart