Arthur™ Image Based Cell Analyzer, NanoEnTek
Supplier: NanoEnTek
AT1000
75794-816EA 7256.25 USD
75794-816
Arthur™ Image Based Cell Analyzer, NanoEnTek
Cell Counters and Flow Cytometers Electronic Cell Counters
The Arthur™ analyzer is an image-based, benchtop cell analyzer designed for fast, accurate cell counting and analysis.
  • Cell counting and viability
  • GFP/RFP expression
  • Annexin V apoptosis analysis
  • Cell cycle assay
  • Advanced accuracy
  • Versatility and flexibility
  • High-speed cell counting and analysis
  • Ease of use
It features two fluorescent channels (green and red) as well as bright field, allowing users to perform a broad range of assays.
Ordering information: Includes power supply, USB drive, and Arthur™ cell analysis slide (50 slides).