High-Definition Videoscope Kit, Extech
Supplier: Extech
HDV540
75853-982EA 2207.57 USD
75853-982
High-Definition Videoscope Kit, Extech
Cameras
VideoScope includes a wired handset + 6 mm dia camera with 240° articulating tip for best viewing.
  • Includes wired handset with nondetachable 6 mm
  • Articulating probe tip adjusts up to 240° viewing angle
  • Compact, high-resolution water-proof (IP67) camera probe
  • Camera head with four built-in bright LED lamps and dimmer to illuminate viewed object
  • AV output for viewing images/video on a monitor
  • Record voice-over audio in video recording mode
  • Store more than 20,000 images on 4GB SD card
The HDV540 is a light weight VideoScope which includes a wired handset and 6mm diameter camera with 240° articulating tip for best viewing angle. Easy one-hand operation of the articulating camera tip facilitates viewing in obstructed areas where standard camera tips cannot access. Images and videos are viewed on the large high resolution color display or external monitors and can be stored in SD memory card (included) or a USB flash drive. Featuring a glare free close up field of view, AV output for viewing images/video on a monitor, Record voice-over audio in video recording mode and memory storage of 20,000 images.
Ordering information: Complete with HDV500 videoscope, handset with articulating probe, SD card, 3.7V rechargeable Li-Polymer battery, 100/240V Universal AC adaptor and multiple plug types (US, EU, UK, AUS), USB and AV cables, and hard carrying case.