Screw Caps for Microcentrifuge Tubes, Bio Plas
Supplier: Bio Plas
4217 4224 4220 4221 4218 4222 4223RSB 4215 4219 4223 4216
20170-748PK 152.98 USD
20170-748 20170-758 20170-746 20170-752 20170-741 20170-750 20170-756 20170-744 20170-754 47734-880 20170-742
Screw Caps for Microcentrifuge Tubes, Bio Plas
Closures Test/Sample Tube Closures
High quality
  • Available in 10 colors
  • Secure and leakproof
  • Ideal for freezer storage, boiling, autoclaving, and irradiation
Bio Plas Screw Cap’s square threads won’t strip. Because the threads are positioned on the outside of the tube, they minimize the risk of contaminating samples or personnel. Caps are available in 10 colors.
Made in the USA.
Packaging: Screw caps are packaged 1000 per pack, 10 packs per case.