Resorufin Cellobioside, Marker Gene Technologies, Inc.
M1238
101447-532EA 170.31 USD
101447-532
Resorufin Cellobioside, Marker Gene Technologies, Inc.
Fluorogenic substrate for detection of cellulase activity.
  • Cellulase substrate
  • Releases the highly fluorescent dye resorufin upon enzyme activity
Resorufin cellobioside is a cellulase substrate that releases the highly fluorescent dye resorufin upon enzyme activity. Since resorufin has a low pKa value, enzyme measurements can be obtained at low pH values, similar to physiological pH's found for many cellulases. Cellulase assays are widely used in the biofuels industry.