Z-Tyr(tBu)-OMe ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
(S)-Methyl 2-(((benzyloxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoate
, N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester
0426435GM
100272-922EA
232.77
USD
100272-922
Z-Tyr(tBu)-OMe ≥99%
Z-Tyr(tBu)-OMe
Formula:
C₂₂H₂₇NO₅ MW: 385.46 g/mol |
MDL Number:
MFCD00237370 CAS Number: 5068-29-1 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...