L-Tyrosine benzyl ester ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Tyr-OBzlBenzyl tyrosinate(S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate

04262025GM
100277-006EA 209.23 USD
100277-006
L-Tyrosine benzyl ester ≥99%, crystalline powder
L-Tyrosine benzyl ester

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₁₆H₁₇NO₃
MW: 271.316 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00191034
CAS Number: 42406-77-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/63/85/21286385.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR