N-Carbobenzoxy-D-tyrosine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Z-D-Tyr-OHN-Cbz-D-tyrosine

0426145GM
100272-910EA 88.92 USD
100272-910
N-Carbobenzoxy-D-tyrosine ≥99%
N-Carbobenzoxy-D-tyrosine
Formula: C₁₇H₁₇NO₅
MW: 315.33 g/mol
MDL Number: MFCD00063031
CAS Number: 64205-12-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/12/21286112.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR