3,6,4'-Trihydroxyflavone ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

T405100MG
100268-464EA 58.85 USD
100268-464
3,6,4'-Trihydroxyflavone ≥98%, crystals
3,6,4'-Trihydroxyflavone

  • Greenish yellow crystals
  • Store in a cool, dry place

Formula: C₁₅H₁₀O₅
Melting Pt: 338…340 °C
MDL Number: MFCD00143079
CAS Number: 253195-19-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/41/21285841.jpg https://us.vwr.com/stibo/web/std.lang.all/58/42/21285842.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR