Trigonelline ≥95% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

021523100MG
100268-670EA 261.54 USD
100268-670
Trigonelline ≥95% (by HPLC), powder
Trigonelline

  • Powder
  • Store at 2-8 °C

Formula: C₇H₇NO₂
Melting Pt: ~230 °C (dec.)
MDL Number: MFCD00054262
CAS Number: 535-83-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/67/21285867.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR