5-(3-(Trifluoromethyl)phenyl)oxazole ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

094011250MG
100272-036EA 130 USD
100272-036
5-(3-(Trifluoromethyl)phenyl)oxazole ≥97%
5-(3-(Trifluoromethyl)phenyl)oxazole
Formula: C₁₀H₆F₃NO
MW: 213.1600 g/mol
MDL Number: MFCD00085180
CAS Number: 175205-48-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/60/70/21286070.jpg https://us.vwr.com/stibo/web/std.lang.all/60/69/21286069.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR