1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%
Supplier: INDOFINE CHEMICAL COMPANY MS
0921601GM
100296-932EA
295
USD
100296-932
1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%
1-(4-(Trifluoromethyl)phenyl)imidazole
Formula:
C₁₀H₇F₃N₂ MW: 212.17 g/mol Melting Pt: 71…73 °C |
MDL Number:
MFCD00060493 CAS Number: 25371-98-6 ADR: , |
![]() ![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...