2-(3-Trifluoromethylphenyl)ethylamine ≥97%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
3-(Trifluoromethyl)phenethylamine
2619401GM
100272-252EA
158.23
USD
100272-252
2-(3-Trifluoromethylphenyl)ethylamine ≥97%
2-(3-Trifluoromethylphenyl)ethylamine
Formula:
C₉H₁₀F₃N MW: 189.18 g/mol |
MDL Number:
MFCD00040756 CAS Number: 52516-30-0 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...