α-Methyl-4-(trifluoromethyl)benzylamine ≥98%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

273021GM
100290-012EA 202.69 USD
100290-012
α-Methyl-4-(trifluoromethyl)benzylamine ≥98%, liquid
alpha-Methyl-4-(trifluoromethyl)benzylamine

  • Colorless liquid to light yellow liquid
  • Store in a cool, dry place

Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
MDL Number: MFCD04038878
CAS Number: 15996-84-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/64/21287064.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR