(α,α,α-Trifluoro-3-tolyl)acetic acid ≥97%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
(3-(Trifluoromethyl)phenyl)acetic acid
, 3-(Trifluoromethyl)phenylacetic acid
2158525GM
100285-378EA
111.15
USD
100285-378
(α,α,α-Trifluoro-3-tolyl)acetic acid ≥97%
(α,α,α-Trifluoro-3-tolyl)acetic acid
Formula:
(C₆H₄(CF₃))CH₂COOH MW: 204.15 g/mol Melting Pt: 76…79 °C |
MDL Number:
MFCD00004339 CAS Number: 351-35-9 ADR: , |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...