4-(Trifluoromethoxy)thiophenol ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-(Trifluoromethoxy)benzenethiol4-(Trifluoromethoxy)benzene-1-thiol

0924891GM
100273-188EA 137.31 USD
100273-188
4-(Trifluoromethoxy)thiophenol ≥95%
4-(Trifluoromethoxy)thiophenol
Formula: C₇H₅F₃OS
MW: 194.18 g/mol
MDL Number: MFCD00052312
CAS Number: 169685-29-4
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/61/27/21286127.jpg https://us.vwr.com/stibo/web/std.lang.all/61/28/21286128.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR