3,4,5-Trifluoroanisole

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1,2,3-Trifluoro-5-methoxybenzene1-Methoxy-3,4,5-trifluorobenzene

075365GM
100273-056EA 300.77 USD
100273-056
3,4,5-Trifluoroanisole
3,4,5-Trifluoroanisole
Formula: C₇H₅F₃O
MW: 162.11 g/mol
MDL Number: MFCD01631451
CAS Number: 203245-17-4
UN: 3271
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/61/24/21286124.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR