Triethylsilyl trifluoromethanesulfonate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Triethylsilyl trifluoromethanesulfonate

26182525GM
100286-206EA 122.92 USD
100286-206
Triethylsilyl trifluoromethanesulfonate ≥99%
Triethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
MDL Number: MFCD00000407
CAS Number: 79271-56-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/69/75/21286975.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR