Triethyloxonium tetrafluoroborate ≥95% in dichloromethane
Supplier: INDOFINE CHEMICAL COMPANY MS
26182425GM
100272-236EA
328.23
USD
100272-236
Triethyloxonium tetrafluoroborate ≥95% in dichloromethane
Triethyloxonium tetrafluoroborate
Formula:
C₆H₁₅BF₄O Melting Pt: 84…89 °C |
MDL Number:
MFCD00044423 CAS Number: 368-39-8 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...