Tribehenin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glyceryl behenateGlyceryl tribehenateGlycerol Tridocosanoate1,2,3-Tridocosanoyl Glycerol

332200100MG
100290-468EA 302.08 USD
100290-468
Tribehenin ≥99%
Tribehenin
Formula: C₆₉H₁₃₄O₆
MW: 1059.83 g/mol
MDL Number: MFCD00056274
CAS Number: 18641-57-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/93/21287093.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR