Triarachidonin (5C,8C,11C,14C) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

33204025MG
100278-308EA 451.15 USD
100278-308
Triarachidonin (5C,8C,11C,14C) ≥99%
Triarachidonin (5C,8C,11C,14C)
MDL Number: MFCD00056307
CAS Number: 23314-57-0
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR