L-Threoninamide hydrochloride ≥98%, powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
H-Thr-NH₂·HCl
, (2S,3R)-2-Amino-3-hydroxybutanamide hydrochloride
0424981GM
100280-778EA
138.62
USD
100280-778
L-Threoninamide hydrochloride ≥98%, powder
L-Threoninamide hydrochloride
- White powder
- Store in a cool, dry place
Formula:
C₄H₁₁ClN₂O₂ MW: 154.594 g/mol Melting Pt: 232…240 °C |
MDL Number:
MFCD00058288 CAS Number: 33209-01-7 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...