n-Tetratriacontane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tetratriacontane

053400100MG
100282-188EA 346.54 USD
100282-188
n-Tetratriacontane ≥99%
n-Tetratriacontane
Formula: H₃C(CH₂)₃₂CH₃
MW: 478.93 g/mol
MDL Number: MFCD00009412
CAS Number: 14167-59-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/92/21286792.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR