(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥99%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Tic-OH(R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic AcidD-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

0424771GM
100276-886EA 28.71 USD
100276-886
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥99%, solid
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

  • White yellow solid
  • Store at 0-8 °C

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
MDL Number: MFCD00144038
CAS Number: 103733-65-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/63/79/21286379.jpg https://us.vwr.com/stibo/web/std.lang.all/63/77/21286377.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR