2,3,4,5-Tetrafluorobenzamide

Supplier: Avantor

075225GM
100295-438EA 198 USD
100295-438
2,3,4,5-Tetrafluorobenzamide
2,3,4,5-Tetrafluorobenzamide
Formula: C₇H₃F₄NO
MDL Number: MFCD01631457
CAS Number: 16582-94-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/14/21287414.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR