n-Tetradecane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tetradecane

051400100MG
100274-326EA 115.08 USD
100274-326
n-Tetradecane ≥99%
n-Tetradecane
Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
MDL Number: MFCD00008986
CAS Number: 629-59-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/87/21286187.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR