Nonadecanoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nonadecylic acid

102400100MG
100299-068EA 329.54 USD
100299-068
Nonadecanoic acid ≥99%
Nonadecanoic acid
Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00002754
CAS Number: 646-30-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/37/21287537.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR