α-Terpinene ≥90%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Isopropyl-4-methylcyclohexa-1,3-dienep-Mentha-1,3-diene

025064S1GM
100269-968EA 137.31 USD
100269-968
α-Terpinene ≥90%, liquid
α-Terpinene

  • Liquid
  • Store at 2-8 °C

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001534
CAS Number: 99-86-5
UN: 2319
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/59/39/21285939.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR