Syringic acid ≥99%, Powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
4-Hydroxy-3,5-dimethoxybenzoic acid
02610725GM
100266-460EA
47.08
USD
100266-460
Syringic acid ≥99%, Powder
Syringic acid
- White shining powder
- Store in a cool, dry place
Formula:
C₉H₁₀O₅ MW: 198.18 g/mol Melting Pt: 208…210 °C |
MDL Number:
MFCD00002552 CAS Number: 530-57-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...