Stevioside, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate

02371810MG
100268-756EA 231.46 USD
100268-756
Stevioside, powder
Stevioside

  • White fluffy powder
  • Store at 2-8 °C

Formula: C₃₈H₆₀O₁₈
MW: 804.88 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD00079561
CAS Number: 57817-89-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/70/21285870.jpg https://us.vwr.com/stibo/web/std.lang.all/58/71/21285871.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR