Pseudochelerythrine nitrate ≥95% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02182710MG
100263-018EA 392.31 USD
100263-018
Pseudochelerythrine nitrate ≥95% (by HPLC), powder
Pseudochelerythrine nitrate

  • Powder
  • Store in a cool, dry place

Formula: C₂₀H₁₄N₂O₇
MDL Number: MFCD00189438
CAS Number: 4752-86-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/28/21285528.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR