Quercetin-3-O-glucose-6"-acetate ≥85% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02109910MG
100266-056EA 449.85 USD
100266-056
Quercetin-3-O-glucose-6"-acetate ≥85% (by HPLC), powder
Quercetin-3-O-glucose-6"-acetate

  • Yellow powder
  • Store in a cool, dry place

Formula: C₂₃H₂₂O₁₃
MDL Number: MFCD00145197
CAS Number: 54542-51-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/97/21285697.jpg https://us.vwr.com/stibo/web/std.lang.all/56/96/21285696.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR