3,5,7,3',4'-Pentahydroxyflavone ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

021135S10MG
100268-640EA 163.46 USD
100268-640
3,5,7,3',4'-Pentahydroxyflavone ≥99%, powder
3,5,7,3',4'-Pentahydroxyflavone

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₅H₁₀O₇
MW: 302.25 g/mol
Melting Pt: 305…312 °C
MDL Number: MFCD00149487
CAS Number: 117-39-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/62/21285862.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR