Fumarine hydrochloride ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

02188010MG
100266-130EA 521.77 USD
100266-130
Fumarine hydrochloride ≥95% (by HPLC)
Fumarine hydrochloride
Formula: C₂₀H₁₉NO₅·HCl
MDL Number: MFCD00236406
CAS Number: 6164-47-2
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR