D-Proline methyl ester hydrochloride ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Pro-OMe.HCl

0423691GM
100280-596EA 104.62 USD
100280-596
D-Proline methyl ester hydrochloride ≥99%
D-Proline methyl ester hydrochloride
Formula: C₆H₁₁NO₂
MW: 165.62 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD00083685
CAS Number: 65365-28-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/38/21286638.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR