D-Proline ethyl ester hydrochloride ≥97%, powder

Supplier: Avantor

Synonyms: H-D-Pro-OEt hydrochloride

0417525GM
100279-734EA 135 USD
100279-734
D-Proline ethyl ester hydrochloride ≥97%, powder
D-Proline ethyl ester hydrochloride

  • White powder
  • Store at 0-8 °C

Formula: C₇H₁₄ClNO₂
MW: 179.644 g/mol
MDL Number: MFCD00191003
CAS Number: 131477-20-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/73/51/21287351.jpg https://us.vwr.com/stibo/web/std.lang.all/73/52/21287352.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR