2-Phenyl butyramide ≥98%, crystalline powder

Supplier: Avantor

240025GM
100269-518EA 28 USD
100269-518
2-Phenyl butyramide ≥98%, crystalline powder
2-Phenyl butyramide

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₁₀H₁₃NO
Melting Pt: 83…85 °C
MDL Number: MFCD00025511
CAS Number: 90-26-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/00/21285900.jpg https://us.vwr.com/stibo/web/std.lang.all/59/01/21285901.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR