1H,1H,2H,2H-Heneicosafluoro-1-dodecanol ≥97%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanol1H,1H,2H,2H-Perfluoro-1-dodecanol1H,1H,2H,2H-Perfluorododecan-1-ol

0918955GM
100273-150EA 215.77 USD
100273-150
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol ≥97%, solid
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol

  • White,waxy solid
  • Store in a cool, dry place

Formula: C₁₂H₅F₂₁O
MW: 564,14 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00039545
CAS Number: 865-86-1
EINECS: 212-748-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/25/21286125.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR