5,7,3',4',5'-Pentamethoxyflavone ≥97%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

P20210MG
100268-458EA 111.15 USD
100268-458
5,7,3',4',5'-Pentamethoxyflavone ≥97%, crystals
5,7,3',4',5'-Pentamethoxyflavone

  • White crystals
  • Store in a cool, dry place

Formula: C₂₀H₂₀O₇
Melting Pt: 190…192 °C
MDL Number: MFCD00017634
CAS Number: 53350-26-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/40/21285840.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR